Learn More
Medchemexpress LLC Tromantadine hydrochloride | 41544-24-5 | 316.87 | 1 ML
SDP
Catalog No. 5000448496
Encompass_Preferred
Tromantadine hydrochloride is an Amantadine derivative that exhibits antiherpetic activity by inhibiting the replication of herpes simplex virus type 1 (HSV-1) and HSV-2. It is intended for research use only.
  • Antiherpetic activity
  • Inhibits herpes simplex virus type 1 (HSV-1) and HSV-2 replication
  • Reduces HSV-induced cytopathic effect
  • Inhibits virus production
Catalog No. 50-004-48496 Supplier Medchemexpress LLC Supplier No. HYU00124B10MM/1ML
Tromantadine hydrochloride is an Amantadine derivative that exhibits antiherpetic activity by inhibiting the replication of herpes simplex virus type 1 (HSV-1) and HSV-2. It is intended for research use only.
  • Antiherpetic activity
  • Inhibits herpes simplex virus type 1 (HSV-1) and HSV-2 replication
  • Reduces HSV-induced cytopathic effect
  • Inhibits virus production