Learn More
Medchemexpress LLC Trometamol acetate | 6850-28-8 | 99.0% | 50 G
SDP
Catalog No. 5000411060
Encompass_Preferred
THAM acetate (Tris acetate) is a biologically inert amino alcohol with low toxicity that buffers carbon dioxide and acids both in vitro and in vivo. It is an effective amine compound for controlling pH within the physiological range.
  • Biologically inert amino alcohol
  • Low toxicity
  • Buffers carbon dioxide and acids in vitro and in vivo
  • Effective for pH control in the physiological range
Catalog No. 50-004-11060 Supplier Medchemexpress LLC Supplier No. HYD0227B50G
THAM acetate (Tris acetate) is a biologically inert amino alcohol with low toxicity that buffers carbon dioxide and acids both in vitro and in vivo. It is an effective amine compound for controlling pH within the physiological range.
  • Biologically inert amino alcohol
  • Low toxicity
  • Buffers carbon dioxide and acids in vitro and in vivo
  • Effective for pH control in the physiological range