Learn More
Selleck Chemical LLC Trometamol hydrochloride S5300-100mg
SDP
Catalog No. 503112297
Encompass_Preferred
Trometamol (THAM Tromethamine TRIS-HCL) hydrochloride a proton acceptor is a biologically inert amino alcohol of low toxicity which buffers carbon dioxide and acids in vitro and in vivo Trometamol rapidly restores pH and acid-base regulation in acidaemia
Catalog No. 50-311-2297 Supplier Selleck Chemical LLC Supplier No. S5300100MG
Trometamol (THAM Tromethamine TRIS-HCL) hydrochloride a proton acceptor is a biologically inert amino alcohol of low toxicity which buffers carbon dioxide and acids in vitro and in vivo Trometamol rapidly restores pH and acid-base regulation in acidaemia