Learn More
U.S. Pharmacopeia Tromethamine, 77-86-1, MFCD00004679, 125mg
Catalog No. NC9296742
Encompass
Molecular formula C4H11NO3, Molecular weight 121.14, Melting Point 339.8 - 341.6 °F (171 - 172 °C), Boiling Point 426.2 - 428 °F (219 - 220 °C), Synonyms: Trometamol, TRIS
Catalog No. NC9296742 Supplier U.S. Pharmacopeia Supplier No. 1698002
Molecular formula C4H11NO3, Molecular weight 121.14, Melting Point 339.8 - 341.6 °F (171 - 172 °C), Boiling Point 426.2 - 428 °F (219 - 220 °C), Synonyms: Trometamol, TRIS