Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tromethamine, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18607609 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-609 125 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-609 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR149125GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TR149, 77-86-1, C4H11NO3

Spectrum™ Chemical Tromethamine, USP is used to treat metabolic acidosis. All Spectrum Chemical USP products, including all USP tromethamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO

Specifications

CAS Max % 1
pH 10.0 to 11.5
Ignition Residue 0.001
Loss on Drying 0.01
Melting Range 168-172°C
Packaging Poly Bottle
Quantity 125 g
Percent Purity 99 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Tromethamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.