Learn More
ACROBiosystems ACROBiosystems Biotinylated Human TROP-2 / TACSTD2 Protein, His,Avitag™
Catalog No. 501956929
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human TROP-2, His,Avitag (TR2-H82E5) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 31.6 kDa. The protein migrates as 42-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6929 Supplier ACROBiosystems Supplier No. TR2H82E5200UG
Recombinant Protein;200UG;Biotinylated Human TROP-2, His,Avitag (TR2-H82E5) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 31.6 kDa. The protein migrates as 42-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.