Learn More
Genscript Corporation TROP-2/TACSTD2Biotin His
Catalog No. 5000056675
Encompass
Trop-2 also known as epithelial glycoprotein-1 antigen (EGP-1) is a protein that in humans is encoded by the TACSTD2 gene Mutations of this gene result in gelatinous drop-like corneal dystrophy an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness
Catalog No. 50-000-56675 Supplier Genscript Corporation Supplier No. Z03841500
Trop-2 also known as epithelial glycoprotein-1 antigen (EGP-1) is a protein that in humans is encoded by the TACSTD2 gene Mutations of this gene result in gelatinous drop-like corneal dystrophy an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness