Learn More
Sino Biological TROP2 / TACSTD2 Antibody (Biotin), Mouse MAb 100ug
Catalog No. 502247612
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TROP2 extracellular domain (rh TROP2; Catalog#10428-H08H; NP_002344.2; Met 1-Thr 274) and then biotinylated. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.
This item is not returnable. View return policy
Catalog No. 50-224-7612 Supplier Sino Biological Supplier No. 10428MM01B100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TROP2 extracellular domain (rh TROP2; Catalog#10428-H08H; NP_002344.2; Met 1-Thr 274) and then biotinylated. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.