Learn More
ACROBiosystems Recombinant Protein;Human TROP-2 / TACSTD2 Protein;HEK293;200UG;TR2,TACSTD2,GA733-1,M1S1,TROP2,TACSTD-2,TROP-2
Catalog No. 501658005
Encompass_Preferred
Recombinant Protein;Human TROP-2 / TACSTD2 Protein;200UG;Human TROP-2 (TR2-H5223) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.8 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8005 Supplier ACROBiosystems Supplier No. TR2H5223200UG
Recombinant Protein;Human TROP-2 / TACSTD2 Protein;200UG;Human TROP-2 (TR2-H5223) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.8 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.