Learn More
Tropaeolin O Indicator (Sulfo Orange 0.1% w/v), Reagents
Specifications
Specifications
| Chemical Name or Material | Tropaeolin O Indicator (Sulfo Orange 0.1% w/v) |
| Boiling Point | 100°C |
| CAS | 7732-18-5, 547-57-9 |
| Packaging | Glass Bottle |
| Specific Gravity | 1.0018 |
| Synonym | Sulfo Orange |
| Color | Orange |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.