Learn More
Sciencell Research Laboratories TROPHOBLAST MEDIUM
Catalog No. NC1786333
Encompass
M consists of 500 ml of basal medium, 25 ml of fetal bovine serum (FBS, Cat. No. 0025), 5 ml of trophoblast growth supplement (TGS, Cat. No. 7152) and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).
Catalog No. NC1786333 Supplier Sciencell Research Laboratories Supplier No. 7121
M consists of 500 ml of basal medium, 25 ml of fetal bovine serum (FBS, Cat. No. 0025), 5 ml of trophoblast growth supplement (TGS, Cat. No. 7152) and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).