Learn More
Medchemexpress LLC HY-B0321 100mg Medchemexpress, Tropicamide CAS:1508-75-4 Purity:>98%
SDP
Catalog No. 502082467
Encompass_Preferred
Medchemexpress, HY-B0321 100mg Tropicamide CAS:1508-75-4 Tropicamide (Ro 1-7683) is a selective M4 muscarinic acetylcholine receptor antagonist. Tropicamide produces short acting mydriasis (dilation of the pupil) and cycloplegia when applied as eye drops. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2467 Supplier Medchemexpress LLC Supplier No. HYB0321100MG
Medchemexpress, HY-B0321 100mg Tropicamide CAS:1508-75-4 Tropicamide (Ro 1-7683) is a selective M4 muscarinic acetylcholine receptor antagonist. Tropicamide produces short acting mydriasis (dilation of the pupil) and cycloplegia when applied as eye drops. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.