Learn More
Medchemexpress LLC Tropicamide-d3 | 2673270-13-6 | 99.10% | 1 MG
SDP
Catalog No. 5000406725
Encompass_Preferred
Tropicamide-d3 is a deuterium labeled Tropicamide and a selective M4 muscarinic acetylcholine receptor antagonist. It produces short acting mydriasis (dilation of the pupil) and cycloplegia when applied as eye drops. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06725 Supplier Medchemexpress LLC Supplier No. HYB0321S1MG
Tropicamide-d3 is a deuterium labeled Tropicamide and a selective M4 muscarinic acetylcholine receptor antagonist. It produces short acting mydriasis (dilation of the pupil) and cycloplegia when applied as eye drops. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.