Learn More
TARGETMOL CHEMICALS INC TROPIFEXOR 5MG
SDP
Catalog No. 502579893
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tropifexor (LJN452) is a novel and highly potent agonist of FXR with an EC50 of 0.2 nM. purity: 99%
Catalog No. 50-257-9893 Supplier TARGETMOL CHEMICALS INC Supplier No. T43795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tropifexor (LJN452) is a novel and highly potent agonist of FXR with an EC50 of 0.2 nM. purity: 99%