Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tropine | 120-29-6 | 25 G
SDP

Catalog No. 5000420740
Encompass_Preferred

Tropine is a tropane alkaloid that serves as an important intermediate for the synthesis of anticholinergics and bronchodilators.

  • For research use only
  • Purity of 99.26%
  • Appears as a white to light yellow solid
  • Classified as an alkaloid
  • Derived from plants such as Solanaceae and Przewalskia tangutica Maximo.
  • Targeted as a drug intermediate
  • Soluble in DMSO (100 mg/mL) and H2O (50 mg/mL)

Catalog No. 50-004-20740 Supplier Medchemexpress LLC Supplier No. 25GHYN7061
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tropine is a tropane alkaloid that serves as an important intermediate for the synthesis of anticholinergics and bronchodilators.

  • For research use only
  • Purity of 99.26%
  • Appears as a white to light yellow solid
  • Classified as an alkaloid
  • Derived from plants such as Solanaceae and Przewalskia tangutica Maximo.
  • Targeted as a drug intermediate
  • Soluble in DMSO (100 mg/mL) and H2O (50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.