Learn More
Medchemexpress LLC HY-Y0135 100mg Medchemexpress, Tropinone CAS:532-24-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0135100MG
Encompass_Preferred
Medchemexpress, HY-Y0135 100mg Tropinone CAS:532-24-1 Tropinone, an alkaloid, acts as a synthetic intermediate to Atropine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0754