Learn More
Medchemexpress LLC Tropinone | 532-24-1 | MFCD00005549 | 100.0% | 139.20 | C8H13NO | 250g
SDP
Catalog No. 5000268585
Encompass_Preferred
Tropinone is an organic synthesis intermediate and a skeleton for constructing derivatives containing thiazole rings and other structures Tropinone derivatives have anticancer activities such as inducing tumor cell apoptosis and inhibiting tyrosinase activity and can be used in research such as anticancer drug development and tyrosinase inhibitor design[1]
Catalog No. 50-002-68585 Supplier Medchemexpress LLC Supplier No. HYY0135250G
Tropinone is an organic synthesis intermediate and a skeleton for constructing derivatives containing thiazole rings and other structures Tropinone derivatives have anticancer activities such as inducing tumor cell apoptosis and inhibiting tyrosinase activity and can be used in research such as anticancer drug development and tyrosinase inhibitor design[1]