Learn More
Cayman Chemical TropIsetron-d5 2.5mg
SDP
Catalog No. 502989332
Encompass_Preferred
An internal standard for the quantification of tropisetron by GC- or LC-MS
Catalog No. 50-298-9332 Supplier Cayman Chemical Supplier No. 307362.5 MG
An internal standard for the quantification of tropisetron by GC- or LC-MS