Learn More
Medchemexpress LLC Tropisetron Hydrochloride | 105826-92-4 | 99.97% | C17H21ClN2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB002010MM/1ML
Encompass_Preferred
Tropisetron Hydrochloride is an effective neuroprotective agent that acts as a 5-HT3 receptor antagonist, a calmodulin inhibitor, and an α7-nAChR receptor agonist, with an IC50 of 70.1 nM for the 5-HT3 receptor. It possesses anti-inflammatory properties and immune-regulating functions, effectively alleviating symptoms associated with chemotherapy and post-surgery. It also reduces Ab-induced hippocampal neuroinflammation.
  • Neuroprotective agent
  • 5-HT3 receptor antagonist
  • Calmodulin inhibitor
  • Α7-nAChR receptor agonist
  • Anti-inflammatory properties
  • Immune-regulating functions
  • Alleviates symptoms associated with chemotherapy and post-surgery
  • Reduces Ab-induced hippocampal neuroinflammation
Catalog No. 50-004-32572