Learn More
AdipoGen Tropodithietic acid TDA
SDP
Catalog No. 502562085
Encompass_Preferred
Chemical. CAS 750590-18-2. Formula C8H4O3S2. MW 212.3. Isolated from Roseobacter gallaeciensis. Antibiotic. Isomeric to thiotropocin. Antibacterial. Active against Gram-positive and Gram-negative bacteria. Antifungal and anti-nematodical. Shows antitumor activity. Cytotoxic against selected mammalian neuronal cell lines. Bacterial signal substance.
Catalog No. 50-256-2085 Supplier AdipoGen Supplier No. BVT0152M005
Chemical. CAS 750590-18-2. Formula C8H4O3S2. MW 212.3. Isolated from Roseobacter gallaeciensis. Antibiotic. Isomeric to thiotropocin. Antibacterial. Active against Gram-positive and Gram-negative bacteria. Antifungal and anti-nematodical. Shows antitumor activity. Cytotoxic against selected mammalian neuronal cell lines. Bacterial signal substance.