Learn More
Medchemexpress LLC Tropolone | 533-75-5 | MFCD00004158 | 100 G
SDP
Catalog No. 5000421097
Encompass_Preferred
Tropolone is a seven-membered non-benzenoid aromatic compound and a precursor of many Azulene derivatives. It is a potent mushroom tyrosinase inhibitor with an IC50 value of 0.4 μM. Tropolone exhibits broad anti-viral and anti-fungal activity and shows synergistic effects when co-treated with nucleos(t)ide analog drugs. It is also a promising candidate for research in osteosarcoma.
  • Potent mushroom tyrosinase inhibitor with an IC50 of 0.4 μM
  • Exhibits broad anti-viral and anti-fungal activity
  • Precursor to Azulene derivatives
  • Promising candidate for osteosarcoma research
  • Solid appearance, off-white to light brown color
  • Purity of 99.77%
Catalog No. 50-004-21097 Supplier Medchemexpress LLC Supplier No. HYN7135100G
Tropolone is a seven-membered non-benzenoid aromatic compound and a precursor of many Azulene derivatives. It is a potent mushroom tyrosinase inhibitor with an IC50 value of 0.4 μM. Tropolone exhibits broad anti-viral and anti-fungal activity and shows synergistic effects when co-treated with nucleos(t)ide analog drugs. It is also a promising candidate for research in osteosarcoma.
  • Potent mushroom tyrosinase inhibitor with an IC50 of 0.4 μM
  • Exhibits broad anti-viral and anti-fungal activity
  • Precursor to Azulene derivatives
  • Promising candidate for osteosarcoma research
  • Solid appearance, off-white to light brown color
  • Purity of 99.77%