Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropolone, G-Biosciences
SCI_ED

Catalog No. S139742
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139742 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139742 Supplier G-Biosciences Supplier No. RC1126
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tropolone is a bactericidal antibiotic composed of a seven-membered aromatic ring. It targets a wide variety of gram-positive and gram-negative bacteria.

Tropolone has been used to prepared fused heterocycles and Ga(III) and In(III) complexes. It is also a reagent used to reduce sugars.

Chemical Identifiers

Molecular Weight (g/mol) 122.12

Specifications

Physical Form Powder
Quantity 5 g
Product Line Gibco
Boiling Point 115°C
Percent Purity 0.98
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.