Learn More
Drg International Inc Troponin I Elisa Ruo
Supplier:  Drg International Inc EIA2952R
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of caridac specific troponin I in serum. This test is intended for the use with serum. This product is for Research Use Only. The cTnI ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay. The assay system utilizes four unique monoclonal antibodies directed against distinct antigenic determinants on the molecule.
Catalog No. 50-194-7551