Learn More
Carbosynth LLC. Purified native Human Troponin I protein (Cardiac)
SDP
Supplier:  Carbosynth LLC. 30AT430.1MG
Encompass_Preferred
Troponin I is a part of the troponin complex. It binds to actin in thin myofilaments to hold the actin-tropomyosin complex in place. Because of it myosin cannot bind actin in relaxed muscle. When calcium binds to the Troponin C it causes conformational changes which lead to dislocation of troponin I and finally tropomyosin leaves the binding site for myosin on actin leading to contraction of muscle. The letter I is given due to its inhibitory character.
Catalog No. 50-125-3090