Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Troponin T (TNNT1) ELISA Kit

Catalog No. EHTNNT1X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHTNNT1X5 5 x 96 Tests
EHTNNT1 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHTNNT1X5 Supplier Invitrogen™ Supplier No. EHTNNT1X5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Troponin T (TNNT1) ELISA quantitates Hu TNNT1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TNNT1. Principle of the method The Human TNNT1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a protein that is a subunit of troponin, which is a regulatory complex located on the thin filament of the sarcomere. This complex regulates striated muscle contraction in response to fluctuations in intracellular calcium concentration. This complex is composed of three subunits: troponin C, which binds calcium, troponin T, which binds tropomyosin, and troponin I, which is an inhibitory subunit. This protein is the slow skeletal troponin T subunit. Mutations in this gene cause nemaline myopathy type 5, also known as Amish nemaline myopathy, a neuromuscular disorder characterized by muscle weakness and rod-shaped, or nemaline, inclusions in skeletal muscle fibers which affects infants, resulting in death due to respiratory insufficiency, usually in the second year. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13805
Assay Range 0.391-25 ng/mL
Assay Sensitivity 0.35 ng/mL
Calibration Range 0.35 ng/mL to 25 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM-9, Angiopoietin 2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGF R, IFN-gamma, IGF-2, IGF-2 R, IL-1R6, IL-24, IL-33, Kallikrein 14, Legumain, LOX-1, MBL, Neprilysin, Notch-1, NOV, Osteoactivin, PD-1, PGRP-5, Serpin A4, sFRP-3, Thrombomodulin, TLR2, TRAIL R1, WIF-1
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ANM; AW146156; Fang2; MGC104241; NEM5; skeletal muscle slow-twitch TnT; slow skeletal muscle troponin T; ssTnT; sTnT; Tnnt1; TNT; TNTS; troponin T slow skeletal muscle type; troponin T slow type isoform sTnT1; troponin T slow type isoform sTnT2; troponin T type 1 (skeletal, slow); troponin T, slow skeletal muscle; troponin T1, skeletal, slow; troponin T1, slow skeletal type; troponin-T1, skeletal, slow; zgc:193831; zgc:193865ANM; AW146156; Fang2; MGC104241; NEM5; skeletal muscle slow-twitch TnT; slow skeletal muscle troponin T; ssTnT; sTnT; Tnnt1; TNT; TNTS; troponin T slow skeletal muscle type; troponin T slow type isoform sTnT1; troponin T slow type isoform sTnT2; troponin T type 1 (skeletal, slow); troponin T, slow skeletal muscle; troponin T1, skeletal, slow; troponin T1, slow skeletal type; troponin-T1, skeletal, slow; zgc:193831; zgc:193865; ANM; NEM5; STNT; TNT; TNTS
Gene ID (Entrez) 7138
Gene Symbol TNNT1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases TnTs, Slow skeletal muscle troponin T, sTnT
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Cardiovascular Research
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym ANM, NEM5, STNT, TNT, TNTS
Target TNNT1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.