Learn More
Medchemexpress LLC HY-B0461 100mg Medchemexpress, Trospium (chloride) CAS:10405-02-4 Purity:>98%
SDP
Catalog No. 502082548
Encompass_Preferred
Medchemexpress, HY-B0461 100mg Trospium (chloride) CAS:10405-02-4 Trospium chloride is a specific and competitive antagonist of muscarinic cholinergic receptors (mAChRs), with antimuscarinic activity. Trospium chloride binds to muscarinic receptors M1, M2 and M3 with high affinity, but not nicotinic, cholinergic receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2548 Supplier Medchemexpress LLC Supplier No. HYB0461100MG
Medchemexpress, HY-B0461 100mg Trospium (chloride) CAS:10405-02-4 Trospium chloride is a specific and competitive antagonist of muscarinic cholinergic receptors (mAChRs), with antimuscarinic activity. Trospium chloride binds to muscarinic receptors M1, M2 and M3 with high affinity, but not nicotinic, cholinergic receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.