Learn More
Apexbio Technology LLC Trospium chloride 10405-02-4 200mg
SDP
Catalog No. 5000090596
Encompass_Preferred
Trospium chloride (CAS 10405-02-4) is a quaternary ammonium compound that functions as a competitive antagonist at muscarinic acetylcholine receptors thereby inhibiting cholinergic stimulation of the bladder detrusor muscle This action reduces involuntary bladder contractions leading to decreased urinary urgency frequency and urge incontinence Trospium chloride displays pharmacological properties distinct from tertiary amine antimuscarinics including limited central nervous system penetration It is widely utilized in preclinical and clinical research focused on urinary disorders and muscarinic receptor pharmacology
Catalog No. 50-000-90596 Supplier Apexbio Technology LLC Supplier No. B1616200
Trospium chloride (CAS 10405-02-4) is a quaternary ammonium compound that functions as a competitive antagonist at muscarinic acetylcholine receptors thereby inhibiting cholinergic stimulation of the bladder detrusor muscle This action reduces involuntary bladder contractions leading to decreased urinary urgency frequency and urge incontinence Trospium chloride displays pharmacological properties distinct from tertiary amine antimuscarinics including limited central nervous system penetration It is widely utilized in preclinical and clinical research focused on urinary disorders and muscarinic receptor pharmacology