Learn More
Cayman Chemical Trotabresib, Purity: ≥98%, CAS: 1706738-98-8, 10 mg
SDP
Catalog No. 502912991
Encompass_Preferred
A BRD4 inhibitor (IC50 = =0.5 µM); inhibits the proliferation of Raji and HL-60 cells (IC50 = =0.5 µM for both)
Catalog No. 50-291-2991 Supplier Cayman Chemical Supplier No. 3629210 MG
A BRD4 inhibitor (IC50 = =0.5 µM); inhibits the proliferation of Raji and HL-60 cells (IC50 = =0.5 µM for both)