Learn More
Medchemexpress LLC Trotabresib | 1706738-98-8 | 99.7% | 383.46 | 100 MG
SDP
Catalog No. 5000351594
Encompass_Preferred
Trotabresib is a reversible and orally active BET inhibitor used in the study of advanced solid tumors. It inhibits the proliferation of diffuse midline glioma (DMG) cells and shows stronger antitumor activity against H3K27M mutant DMG cells than DMG wildtype.
  • Reversible and orally active BET inhibitor
  • Applied in studies for advanced solid tumors
  • Inhibits proliferation of diffuse midline glioma (DMG) cells
  • Shows stronger antitumor activity against H3K27M mutant DMG cells than DMG wildtype
Catalog No. 50-003-51594 Supplier Medchemexpress LLC Supplier No. HY137573100MG
Trotabresib is a reversible and orally active BET inhibitor used in the study of advanced solid tumors. It inhibits the proliferation of diffuse midline glioma (DMG) cells and shows stronger antitumor activity against H3K27M mutant DMG cells than DMG wildtype.
  • Reversible and orally active BET inhibitor
  • Applied in studies for advanced solid tumors
  • Inhibits proliferation of diffuse midline glioma (DMG) cells
  • Shows stronger antitumor activity against H3K27M mutant DMG cells than DMG wildtype