Learn More
Medchemexpress LLC Trotabresib | 1706738-98-8 | 99.7% | 383.46 | 50 MG
SDP
Catalog No. 5000348045
Encompass_Preferred
Trotabresib (CC-90010) is a reversible and orally active BET inhibitor. It is used in the study of advanced solid tumors and inhibits the proliferation of diffuse midline glioma (DMG) cells.
  • Reversible and orally active BET inhibitor
  • Applied in the study for advanced solid tumors
  • Inhibits proliferation of diffuse midline glioma (DMG) cells
  • Exhibits stronger antitumor activity against H3K27M mutant DMG cells than DMG wildtype
Catalog No. 50-003-48045 Supplier Medchemexpress LLC Supplier No. HY13757350MG
Trotabresib (CC-90010) is a reversible and orally active BET inhibitor. It is used in the study of advanced solid tumors and inhibits the proliferation of diffuse midline glioma (DMG) cells.
  • Reversible and orally active BET inhibitor
  • Applied in the study for advanced solid tumors
  • Inhibits proliferation of diffuse midline glioma (DMG) cells
  • Exhibits stronger antitumor activity against H3K27M mutant DMG cells than DMG wildtype