Learn More
Lonza Walkersville Lonza Trough, Disposable, 100 μL, Endotoxin-Free
Supplier:  Lonza Walkersville 00229888
Encompass
This disposable trough is designed for use with the PyroTec™ PRO Robotic Solution. It is a consumable item essential for automated endotoxin detection processes, ensuring reliable results.
  • Certified to be endotoxin-free (<0.005 EU/mL).
  • Used with the PyroTec™ PRO Robotic System.
  • Part of a suite of consumables for automated, plate-based endotoxin detection.
  • Has a 100 μL capacity.
Catalog No. NC3986792