Learn More
Biosure Inc TROUT ERYTHROCYTE NUCLEI, 2ML
Catalog No. NC0484690
Encompass
BioSure Trout Erythrocyte Nuclei (TEN's) are an internal biological reference material that can function as a DNA standard when determining nuclear DNA content by flow cytometry.
Catalog No. NC0484690 Supplier Biosure Inc Supplier No. 1007
BioSure Trout Erythrocyte Nuclei (TEN's) are an internal biological reference material that can function as a DNA standard when determining nuclear DNA content by flow cytometry.