Learn More
World Precision Instrument Troutman Tying Forceps, 10 cm, Curved
SDP
Supplier:  World Precision Instrument WP505210
Encompass
  • Thumb Forceps
  • 10cm (3.94")
  • 0.52mm fine tip
  • Curved
  • Stainless Steel
Catalog No. 50-253-8291