Learn More
TARGETMOL CHEMICALS INC Trovafloxacin mesylate 10MG
SDP
Catalog No. 502876133
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trovafloxacin is a broad-spectrum fluoroquinolone antibiotic that inhibits the activity of DNA gyrase and topoisomerase IV, enzymes involved in DNA replication. It also acts as an effective and specific inhibitor of the pannexin 1 channel (PANX1, IC50 = 4 uM). Purity 99.12%
Catalog No. 50-287-6133 Supplier TARGETMOL CHEMICALS INC Supplier No. T13231L10MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trovafloxacin is a broad-spectrum fluoroquinolone antibiotic that inhibits the activity of DNA gyrase and topoisomerase IV, enzymes involved in DNA replication. It also acts as an effective and specific inhibitor of the pannexin 1 channel (PANX1, IC50 = 4 uM). Purity 99.12%