Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trovafloxacin mesylate | 147059-75-4 | MFCD00913361 | 512.46 g·mol-1 | C21H19F3N4O6S | 5 MG
SDP

Catalog No. 5000246776
Encompass_Preferred

Trovafloxacin mesylate is a fluoroquinolone-class antibiotic supplied as the mesylate salt for research use. It is intended for laboratory studies involving antimicrobial testing and investigations of DNA gyrase and topoisomerase IV inhibition.

  • broad-spectrum fluoroquinolone antibiotic for research applications.
  • CAS number 147059-75-4 for unambiguous identification.
  • molecular formula C21H19F3N4O6S and molecular weight 512.46 g·mol-1.
  • soluble in DMSO (≈116.67 mg/mL) and water (≈20 mg/mL) with ultrasonic assistance.
  • formulatable for in vivo use at ≥2.08 mg/mL in recommended vehicle systems.
  • available in small pack sizes suitable for laboratory experiments (1 mg, 5 mg, 10 mg).

Catalog No. 50-002-46776 Supplier Medchemexpress LLC Supplier No. HY1033995MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trovafloxacin mesylate is a fluoroquinolone-class antibiotic supplied as the mesylate salt for research use. It is intended for laboratory studies involving antimicrobial testing and investigations of DNA gyrase and topoisomerase IV inhibition.

  • broad-spectrum fluoroquinolone antibiotic for research applications.
  • CAS number 147059-75-4 for unambiguous identification.
  • molecular formula C21H19F3N4O6S and molecular weight 512.46 g·mol-1.
  • soluble in DMSO (≈116.67 mg/mL) and water (≈20 mg/mL) with ultrasonic assistance.
  • formulatable for in vivo use at ≥2.08 mg/mL in recommended vehicle systems.
  • available in small pack sizes suitable for laboratory experiments (1 mg, 5 mg, 10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.