Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trovafloxacin mesylate | 147059-75-4 | MFCD00913361 | 95.0% | 512.46 g/mol | C21H19F3N4O6S | 10 MG
SDP

Catalog No. 5000246898
Encompass_Preferred

Trovafloxacin mesylate is the mesylate salt of trovafloxacin, a broad-spectrum fluoroquinolone antibiotic used as a research reagent. It inhibits bacterial DNA gyrase and topoisomerase IV and is supplied in low-milligram amounts with solubility and storage guidance for laboratory use.

  • Chemical formula: C21H19F3N4O6S.
  • Molecular weight: 512.46 g/mol.
  • CAS number: 147059-75-4.
  • Typical package amount: 10 mg.
  • Solubility: DMSO up to 116.67 mg/mL; water up to 20 mg/mL (may need ultrasonic).
  • Storage: store under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Applications: in vitro and in vivo research, enzyme inhibition assays.

Catalog No. 50-002-46898 Supplier Medchemexpress LLC Supplier No. HY10339910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trovafloxacin mesylate is the mesylate salt of trovafloxacin, a broad-spectrum fluoroquinolone antibiotic used as a research reagent. It inhibits bacterial DNA gyrase and topoisomerase IV and is supplied in low-milligram amounts with solubility and storage guidance for laboratory use.

  • Chemical formula: C21H19F3N4O6S.
  • Molecular weight: 512.46 g/mol.
  • CAS number: 147059-75-4.
  • Typical package amount: 10 mg.
  • Solubility: DMSO up to 116.67 mg/mL; water up to 20 mg/mL (may need ultrasonic).
  • Storage: store under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Applications: in vitro and in vivo research, enzyme inhibition assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.