Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trovafloxacin (mesylate) | 147059-75-4 | MFCD00913361 | 99.6% | 512.46 g·mol⁻¹ | C21H19F3N4O6S | 100 MG
SDP

Catalog No. 5000246799
Encompass_Preferred

Trovafloxacin mesylate is the methanesulfonate (mesylate) salt of trovafloxacin, a fluoroquinolone antibiotic used as a research tool compound and analytical reference in microbiological and biochemical studies.

  • Fluoroquinolone antibiotic salt suitable for research and analytical applications.
  • Broad-spectrum activity against Gram-positive, Gram-negative, and anaerobic bacteria.
  • Molecular formula C21H19F3N4O6S and molecular weight 512.46 g·mol⁻¹.
  • CAS number 147059-75-4 for precise identification.
  • Soluble in DMSO; commonly supplied as a dry powder for laboratory use.

Catalog No. 50-002-46799 Supplier Medchemexpress LLC Supplier No. HY103399100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trovafloxacin mesylate is the methanesulfonate (mesylate) salt of trovafloxacin, a fluoroquinolone antibiotic used as a research tool compound and analytical reference in microbiological and biochemical studies.

  • Fluoroquinolone antibiotic salt suitable for research and analytical applications.
  • Broad-spectrum activity against Gram-positive, Gram-negative, and anaerobic bacteria.
  • Molecular formula C21H19F3N4O6S and molecular weight 512.46 g·mol⁻¹.
  • CAS number 147059-75-4 for precise identification.
  • Soluble in DMSO; commonly supplied as a dry powder for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.