Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trovafloxacin mesylate | 147059-75-4 | MFCD00913361 | 99.6% | 512.46 g·mol⁻¹ | C21H19F3N4O6S | 50 MG
SDP

Catalog No. 5000244777
Encompass_Preferred

Trovafloxacin mesylate is the methanesulfonate (mesylate) salt of trovafloxacin, a broad-spectrum fluoroquinolone antibiotic supplied for research use. It inhibits bacterial DNA gyrase and topoisomerase IV and has been reported to block pannexin 1 (PANX1) channels in cellular assays.

  • Broad-spectrum fluoroquinolone antibiotic and research reagent.
  • Inhibits DNA gyrase and topoisomerase IV.
  • Reported PANX1 inhibitor in cellular assays.
  • Molecular formula C21H19F3N4O6S; molecular weight 512.46 g·mol⁻¹.
  • Salt form: mesylate (methanesulfonate).
  • Packaged as 50 mg for research use.
  • CAS number 147059-75-4.

Catalog No. 50-002-44777 Supplier Medchemexpress LLC Supplier No. HY10339950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trovafloxacin mesylate is the methanesulfonate (mesylate) salt of trovafloxacin, a broad-spectrum fluoroquinolone antibiotic supplied for research use. It inhibits bacterial DNA gyrase and topoisomerase IV and has been reported to block pannexin 1 (PANX1) channels in cellular assays.

  • Broad-spectrum fluoroquinolone antibiotic and research reagent.
  • Inhibits DNA gyrase and topoisomerase IV.
  • Reported PANX1 inhibitor in cellular assays.
  • Molecular formula C21H19F3N4O6S; molecular weight 512.46 g·mol⁻¹.
  • Salt form: mesylate (methanesulfonate).
  • Packaged as 50 mg for research use.
  • CAS number 147059-75-4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.