Learn More
Medchemexpress LLC Trovafloxacin mesylate | 147059-75-4 | MFCD00913361 | 99.8% | 512.46 g/mol | C21H19F3N4O6S | 1 ML
SDP
Catalog No. 5000246489
Encompass_Preferred
Trovafloxacin mesylate is the mesylate salt of trovafloxacin, a broad-spectrum fluoroquinolone antibiotic that inhibits DNA gyrase and topoisomerase IV. It is used in research to study antibacterial activity and pannexin 1 (PANX1) channel function and inhibition.
  • Broad-spectrum fluoroquinolone antibiotic that inhibits DNA gyrase and topoisomerase IV.
  • Also reported as a pannexin 1 (PANX1) inhibitor with an IC50 around 4 μM.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for experimental flexibility.
  • High purity: 99.8% (HPLC) as reported by supplier.
  • CAS number 147059-75-4; molecular weight 512.46 g/mol.
Catalog No. 50-002-46489 Supplier Medchemexpress LLC Supplier No. HY10339910MM 1ML
Trovafloxacin mesylate is the mesylate salt of trovafloxacin, a broad-spectrum fluoroquinolone antibiotic that inhibits DNA gyrase and topoisomerase IV. It is used in research to study antibacterial activity and pannexin 1 (PANX1) channel function and inhibition.
  • Broad-spectrum fluoroquinolone antibiotic that inhibits DNA gyrase and topoisomerase IV.
  • Also reported as a pannexin 1 (PANX1) inhibitor with an IC50 around 4 μM.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for experimental flexibility.
  • High purity: 99.8% (HPLC) as reported by supplier.
  • CAS number 147059-75-4; molecular weight 512.46 g/mol.