Learn More
Medchemexpress LLC Trovafloxacin mesylate | 147059-75-4 | MFCD00913361 | 99.8% | 512.46 g/mol | C21H19F3N4O6S | 25 MG
SDP
Catalog No. 5000247230
Encompass_Preferred
Trovafloxacin mesylate is a broad-spectrum fluoroquinolone antibiotic and selective pannexin 1 (PANX1) inhibitor supplied as a high-purity solid for research use. It inhibits bacterial DNA gyrase and topoisomerase IV and blocks PANX1 inward currents (IC50 ≈ 4 μM), making it suitable for antibacterial mechanism and ion channel studies.
  • Broad-spectrum antibacterial activity against Gram-positive, Gram-negative, and anaerobic organisms.
  • Inhibits DNA gyrase and topoisomerase IV to block bacterial replication.
  • Selective PANX1 inhibition useful for ion channel and apoptotic fragmentation studies.
  • High stated purity for consistent experimental results.
  • Stable solid form with recommended storage under nitrogen at -20°C.
Catalog No. 50-002-47230 Supplier Medchemexpress LLC Supplier No. HY10339925MG
Trovafloxacin mesylate is a broad-spectrum fluoroquinolone antibiotic and selective pannexin 1 (PANX1) inhibitor supplied as a high-purity solid for research use. It inhibits bacterial DNA gyrase and topoisomerase IV and blocks PANX1 inward currents (IC50 ≈ 4 μM), making it suitable for antibacterial mechanism and ion channel studies.
  • Broad-spectrum antibacterial activity against Gram-positive, Gram-negative, and anaerobic organisms.
  • Inhibits DNA gyrase and topoisomerase IV to block bacterial replication.
  • Selective PANX1 inhibition useful for ion channel and apoptotic fragmentation studies.
  • High stated purity for consistent experimental results.
  • Stable solid form with recommended storage under nitrogen at -20°C.