Learn More
Selleck Chemical LLC Trovafloxacin Mesylate-5mg
SDP
Catalog No. 502389737
Encompass_Preferred
Trovafloxacin mesylate, a broad-spectrum quinolone antibiotic, is a potent, selective and orally active pannexin 1 channel (PANX1) inhibitor with an IC50 of 4 μM for PANX1 inward current.
Catalog No. 50-238-9737 Supplier Selleck Chemical LLC Supplier No. E28715MG
Trovafloxacin mesylate, a broad-spectrum quinolone antibiotic, is a potent, selective and orally active pannexin 1 channel (PANX1) inhibitor with an IC50 of 4 μM for PANX1 inward current.