Learn More
Abcam Trovaflo x acin mesylate, Fluoroquinolone antibiotic agent, 10MG
Catalog No. 502705625
Encompass_Preferred
MW 512.46 g/mol, Purity >98%. Fluoroquinolone antibiotic agent. Topoisomerase IV and DNA gyrase inhibitor. Reversibly inhibits DNA synthesis. Inhibits PANX1 (IC₅₀ = 4 μM). Shows antibiotic effects in vivo (MIC₉₀ = 244 nM, pneumococci).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5625 Supplier Abcam Supplier No. AB14456210MG
MW 512.46 g/mol, Purity >98%. Fluoroquinolone antibiotic agent. Topoisomerase IV and DNA gyrase inhibitor. Reversibly inhibits DNA synthesis. Inhibits PANX1 (IC₅₀ = 4 μM). Shows antibiotic effects in vivo (MIC₉₀ = 244 nM, pneumococci).
The product is subject to the following: Abcam Restricted Use Statement