Learn More
Cayman Chemical TrovafloxacInmesylate 10mg
SDP
Catalog No. 502892653
Encompass_Preferred
A broad spectrum antibiotic that inhibits the uncoiling of supercoiled DNA in bacteria by blocking the activity of topoisomerase IV (IC50 = 3.02 UG/ml) and DNA gyrase (IC50 = 7.13 UG/ml)
Catalog No. 50-289-2653 Supplier Cayman Chemical Supplier No. 900030310 MG
A broad spectrum antibiotic that inhibits the uncoiling of supercoiled DNA in bacteria by blocking the activity of topoisomerase IV (IC50 = 3.02 UG/ml) and DNA gyrase (IC50 = 7.13 UG/ml)