Learn More
Cayman Chemical Trovafloxacin (mesylate), Purity: ≥98%, CAS: 147059-75-4, 50 mg
SDP
Catalog No. 502892713
Encompass_Preferred
A broad spectrum antibiotic that inhibits the uncoiling of supercoiled DNA in bacteria by blocking the activity of topoisomerase IV (IC50 = 3.02 µg/ml) and DNA gyrase (IC50 = 7.13 µg/ml)
Catalog No. 50-289-2713 Supplier Cayman Chemical Supplier No. 900030350 MG
A broad spectrum antibiotic that inhibits the uncoiling of supercoiled DNA in bacteria by blocking the activity of topoisomerase IV (IC50 = 3.02 µg/ml) and DNA gyrase (IC50 = 7.13 µg/ml)