Learn More
Aviva Systems Biology Corp TROVE2 Antibody - N-terminal region (ARP40535_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40535_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TROVE2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TROVE2. Alternate names: RO60, SSA2, RORNPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57511