Learn More
Selleck Chemical LLC Troxerutin S2369-1g
SDP
Catalog No. 503112731
Encompass_Preferred
Troxerutin a natural bioflavonoid isolated from Sophora japonica has been reported to have many benefits and medicinal properties Troxerutin also known as vitamin P4 is a tri-hydroxyethylated derivative of natural bioflavonoid rutins which can inhibit the production of reactive oxygen species (ROS) and depress ER stress-mediated NOD activation
Catalog No. 50-311-2731 Supplier Selleck Chemical LLC Supplier No. S23691G
Troxerutin a natural bioflavonoid isolated from Sophora japonica has been reported to have many benefits and medicinal properties Troxerutin also known as vitamin P4 is a tri-hydroxyethylated derivative of natural bioflavonoid rutins which can inhibit the production of reactive oxygen species (ROS) and depress ER stress-mediated NOD activation