Learn More
Apexbio Technology LLC Troxerutin 7085-55-4 5g
SDP
Catalog No. 5000062686
Encompass_Preferred
Troxerutin (CAS 7085-55-4) is a bioactive small molecule known to exhibit protective effects against gamma radiation (GR)-induced DNA damage It promotes DNA repair mechanisms thereby reducing radiation-related single- and double-strand breaks Experimentally Troxerutin (1 mM) decreased GR-triggered micronucleus formation in human lymphocytes by approximately 41% In mouse models intraperitoneal administration before or after irradiation (up to 175 mg/kg) minimized micronucleus frequency in reticulocytes and mitigated DNA strand breaks in leukocytes and bone marrow cells with dosage-dependent results Its properties make Troxerutin relevant for radiation biology research particularly in genomic stability and radioprotection studies
Catalog No. 50-000-62686 Supplier Apexbio Technology LLC Supplier No. B59885000
Troxerutin (CAS 7085-55-4) is a bioactive small molecule known to exhibit protective effects against gamma radiation (GR)-induced DNA damage It promotes DNA repair mechanisms thereby reducing radiation-related single- and double-strand breaks Experimentally Troxerutin (1 mM) decreased GR-triggered micronucleus formation in human lymphocytes by approximately 41% In mouse models intraperitoneal administration before or after irradiation (up to 175 mg/kg) minimized micronucleus frequency in reticulocytes and mitigated DNA strand breaks in leukocytes and bone marrow cells with dosage-dependent results Its properties make Troxerutin relevant for radiation biology research particularly in genomic stability and radioprotection studies