Learn More
Medchemexpress LLC Troxerutin | 7085-55-4 | 98.0% | 742.68 | 10 G
SDP
Catalog No. 5000430957
Encompass_Preferred
Troxerutin, also known as vitamin P4, is a tri-hydroxyethylated derivative of natural bioflavonoid rutins. It is capable of inhibiting the production of reactive oxygen species (ROS) and depressing ER stress-mediated NOD activation.
  • Inhibits ROS production
  • Depresses ER stress-mediated NOD activation
  • Lowers body weight and improves metabolic parameters
  • Restores glucose intolerance and insulin signaling
  • Attenuates vascular damages
Catalog No. 50-004-30957 Supplier Medchemexpress LLC Supplier No. HYN013910G
Troxerutin, also known as vitamin P4, is a tri-hydroxyethylated derivative of natural bioflavonoid rutins. It is capable of inhibiting the production of reactive oxygen species (ROS) and depressing ER stress-mediated NOD activation.
  • Inhibits ROS production
  • Depresses ER stress-mediated NOD activation
  • Lowers body weight and improves metabolic parameters
  • Restores glucose intolerance and insulin signaling
  • Attenuates vascular damages