Learn More
Medchemexpress LLC Troxipide | 30751-05-4 | MFCD00866876 | 99.6% | 250 MG
SDP
Catalog No. 5000405575
Encompass_Preferred
Troxipide is an orally active defensive factor-enhancing therapeutic agent for gastritis and gastric ulcer (GU). It is a non-antisecretory gastroprotective agent with antiulcer, anti-inflammatory, and mucus-secreting properties.
  • Enhances gastric mucosal blood flow and gastric mucosal defense factors.
  • Promotes tissue repair, blood circulation, metabolism, and gastric ulcer (GU) repair.
  • Exerts inhibitory, therapeutic, and preventive effects specifically in the stomach.
  • Significantly alleviates gastric ulcer (GU).
  • Reduces levels of IL-17, IL-6, TNF-α, IFN-γ, and AP-1 in gastric ulcer models.
Catalog No. 50-004-05575 Supplier Medchemexpress LLC Supplier No. 250MGHYB0758
Troxipide is an orally active defensive factor-enhancing therapeutic agent for gastritis and gastric ulcer (GU). It is a non-antisecretory gastroprotective agent with antiulcer, anti-inflammatory, and mucus-secreting properties.
  • Enhances gastric mucosal blood flow and gastric mucosal defense factors.
  • Promotes tissue repair, blood circulation, metabolism, and gastric ulcer (GU) repair.
  • Exerts inhibitory, therapeutic, and preventive effects specifically in the stomach.
  • Significantly alleviates gastric ulcer (GU).
  • Reduces levels of IL-17, IL-6, TNF-α, IFN-γ, and AP-1 in gastric ulcer models.