Learn More
AnaSpec TRP-2, Tyrosinase-related Protein 2 (180-188), 1 mg
Catalog No. NC1064018
Encompass
This peptide is derived from tyrosinase-related protein 2 (TRP2) residues 180-188. TRP2 belongs to the melanocyte differentiation antigens and has been implicated as a target for immunotherapy of human as well as murine melanoma. Studies show that this TRP2 derived peptide can bind to mouse and human MHC class I molecules. Immunization with TRP2 peptide loaded dendritic cells (DCs) results in effective induction of antitumor immunity.
Catalog No. NC1064018 Supplier AnaSpec Supplier No. AS61058
Add to Cart
Add to Cart
This peptide is derived from tyrosinase-related protein 2 (TRP2) residues 180-188. TRP2 belongs to the melanocyte differentiation antigens and has been implicated as a target for immunotherapy of human as well as murine melanoma. Studies show that this TRP2 derived peptide can bind to mouse and human MHC class I molecules. Immunization with TRP2 peptide loaded dendritic cells (DCs) results in effective induction of antitumor immunity.